Skip to Content
Merck
All Photos(1)

Key Documents

SRP0178

Sigma-Aldrich

Histone H4 full length human

recombinant, expressed in E. coli, ≥80% (SDS-PAGE)

Synonym(s):

HIST2H4A

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€668.00

€668.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
1 MG
€668.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€668.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥80% (SDS-PAGE)

form

aqueous solution

mol wt

12.1 kDa

packaging

pkg of 1 mg

storage condition

avoid repeated freeze/thaw cycles

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... HIST2H4A(8370)

General description

Human Histone 4 (HIST2H4A) (GenBank Accession No. NM_003548), amino acids 2-104 (end) with N-terminal His-tag, MW = 12.1 kDa, expressed in an E. coli expression system.

Application

Suitable substrate for histone methyltransferases.

Biochem/physiol Actions

Histones H2B and H4 are found to become mobilized during an infection with HSV-1 (herpes simplex virus-1) and become available for binding to viral genome.[1]

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 20% glycerol and 3 mM DTT.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing protein to recover full content of the tube. Aliquot protein into single use aliquots. Store remaining undiluted protein in aliquots at -70°C. Note: Protein is very sensitive to freeze/thaw cycles.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Core histones H2B and H4 are mobilized during infection with herpes simplex virus 1.
Conn KL
Journal of Virology, 85, 13234-13252 (2011)
Recognition and classification of histones using support vector machine.
Bhasin M
Journal of Computational Biology : A Journal of Computational Molecular Cell Biology, 13, 102-112 (2006)
Shrika G Harjivan et al.
Molecules (Basel, Switzerland), 26(5) (2021-04-04)
Nevirapine (NVP), a non-nucleoside reverse transcriptase inhibitor widely used in combined antiretroviral therapy and to prevent mother-to-child transmission of the human immunodeficiency virus type 1, is associated with several adverse side effects. Using 12-mesyloxy-nevirapine, a model electrophile of the reactive
The histone variant H3.3 marks active chromatin by replication-independent nucleosome assembly.
Ahmad K and Henikoff S
Molecular Cell, 9(6), 1191-1200 (2002)
A comprehensive view of the epigenetic landscape part I: DNA methylation, passive and active DNA demethylation pathways and histone variants.
Sadakierska-Chudy A
Neurotoxicity Research, 27(1), 84-97 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service