Skip to Content
Merck
All Photos(1)

Key Documents

SRP0164

Sigma-Aldrich

Histone H4 (2-58) human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE)

Synonym(s):

HIST2H4A

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
€356.00

€356.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
500 μG
€356.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

€356.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

aqueous solution

mol wt

32 kDa

packaging

pkg of 500 μg

storage condition

avoid repeated freeze/thaw cycles

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... HIST2H4A(8370)

General description

Human Histone 4 (GenBank Accession No. NM_003548), (2-58) with N-terminal GST-tag, MW = 32 kDa, expressed in an E. coli expression system.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 20% glycerol and 3 mM DTT.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. What is the amino acid sequence of Histone H4 (2-58) human, Product SRP0164?

    1 answer
    1. The sequence for Product SRP0164, Histone H4 (2-58) human is as follows:SGRGKGGKGLGKGGAKRHRKVLRDNIQGITKPAIRRLARRGGVKRISGLIYEETRGV

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service