Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
biological source
human
Quality Segment
recombinant
expressed in E. coli
mol wt
Mw 11 kDa
manufacturer/tradename
Upstate®
technique(s)
activity assay: suitable, western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
General description
Analysis Note
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| biological source human | biological source rabbit | biological source rabbit | biological source rabbit |
| technique(s) activity assay: suitable, western blot: suitable | technique(s) immunocytochemistry: suitable | technique(s) dot blot: suitable, immunoprecipitation (IP): suitable, western blot: suitable | technique(s) ChIP: 1:20-1:100, immunofluorescence: 1:50-1:200, immunohistochemistry: 1:50-1:200, western blot: 1:200-1:2000 |
| recombinant expressed in E. coli | recombinant - | recombinant - | recombinant - |
| mol wt Mw 11 kDa | mol wt - | mol wt - | mol wt 11 |
| UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
| shipped in dry ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 14-697 | 04053252515033 |



