Skip to Content
Merck

Skip To

SML3633

C23.28-TPP

≥98% (HPLC)

Synonym(s):

(6-(4-(2,5-Dimethylphenyl)-4-oxobutanoyloxy)hexyl)triphenylphosphonium bromide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C36H40O3P·Br
CAS Number:
Molecular Weight:
631.58
NACRES:
NA.21
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Bacteria-permeable prodrug inhibitor of IspH
C23.28-TPP is a bacteria-permeable prodrug inhibitor of IspH that is bactericidal to number of multidrug-resistant bacteria including Acinetobacter, Pseudomonas, Klebsiella, Enterobacter, Vibrio, Shigella, Salmonella, Yersinia, Mycobacterium and Bacillus. It stimulates rapid immune response through activation human Vγ9Vδ2 T cells in a humanized mouse model of bacterial infection. C23.28-TPP was able to clear infection by a clinically isolated multidrug-resistant strain of E. aerogenes (UCI 15) in BALBc mice. It is relatively non-toxic to mammalian cells.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters