Skip to Content
Merck
All Photos(1)

Key Documents

SML2342

Sigma-Aldrich

SGC3027N

≥98% (HPLC)

Synonym(s):

5′-S-[4-[N-3-(2,4,5-trimethyl-3,6-dioxo-cyclohexa-1,4-dienyl)-3-methylbutyryl-N-(4′-chloro-biphenyl-3-ylmethyl]amino]butyl]-2′,3′-O-(1-methylethylidene)-5′-thioadenosine, N-(4-((((3aS,4S,6R,6aR)-6-(6-amino-9H-purin-9-yl)-2,2-dimethyltetrahydrofuro[3,4-d][1,3]dioxol-4-yl)methyl)thio)butyl)-N-((4′-chloro-[1,1′-biphenyl]-3-yl)methyl)-3-methyl-3-(2,4,5-trimethyl-3,6-dioxocyclohexa-1,4-dien-1-yl)butanamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€139.00
25 MG
€451.00

€139.00


Estimated to ship on28 May 2025



Select a Size

Change View
5 MG
€139.00
25 MG
€451.00

About This Item

Empirical Formula (Hill Notation):
C44H51ClN6O6S
CAS Number:
Molecular Weight:
827.43
UNSPSC Code:
41121813
NACRES:
NA.77

€139.00


Estimated to ship on28 May 2025


Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC(C=C1)=CC=C1C2=CC(CN(C(CC(C)(C)C(C(C(C)=C3C)=O)=C(C)C3=O)=O)CCCCSC[C@H]4O[C@@H](N(C=N5)C6=C5C(N)=NC=N6)[C@H]7[C@@H]4OC(C)(C)O7)=CC=C2

InChI key

FJBPSCVGHZASPG-LJVHFRCJSA-N

Biochem/physiol Actions

Control probe for PRMT7 inhibitor SGC3027
SGC3027N is the control probe for the PRMT7 inhibitor SGC3027

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service