Skip to Content
Merck
All Photos(1)

Key Documents

539209

Sigma-Aldrich

Protein Arginine N-Methyltransferase Inhibitor, AMI-1

The Protein Arginine N-Methyltransferase Inhibitor, AMI-1 controls the biological activity of Protein Arginine N-Methyltransferase. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Protein Arginine N-Methyltransferase Inhibitor, AMI-1, PRMT Inhibitor I

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€205.00

€205.00


Estimated to ship on31 May 2025



Select a Size

Change View
5 MG
€205.00

About This Item

Empirical Formula (Hill Notation):
C21H12N2Na4O9S2
Molecular Weight:
592.42
UNSPSC Code:
12352202
NACRES:
NA.77

€205.00


Estimated to ship on31 May 2025


Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

solubility

water: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable, symmetrical sulfonated urea compound that acts as a potent, specific and non-AdoMet (S-adenosyl-L-methionine, SAM)-competitive inhibitor of protein arginine N-methyltransferases (PRMTs; IC50 = 8.81 µM for PRMT1 and 3.03 for yeast-RMT1p) with minimal effect on lysine methyltransferases. Inhibits nuclear receptor reporter gene activation in MCF-7 cells, and HIV-1 RT polymerase (IC50 = 5 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Protein arginine N-methyltransferases (PRMTs)
Product does not compete with ATP.
Reversible: no
Target IC50: 8.81 µM for PRMT1 and 3.03 for yeast-RMT1p

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Cheng, D., et al. 2004. J. Biol. Chem.279, 23892.
Skillman, A.G., et al. 2002. Bioorg. Chem.30, 443.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service