Skip to Content
Merck
All Photos(1)

Key Documents

5.06046

Sigma-Aldrich

PRMT Inhibitor VII, DB867

Synonym(s):

PRMT Inhibitor VII, DB867, 5-[5-(4-Carbamimidoyl-phenyl)-furan-2-yl]-pyridine-2-carboxamidine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€168.00

€168.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€168.00

About This Item

Empirical Formula (Hill Notation):
C17H15NO5
CAS Number:
Molecular Weight:
313.30
UNSPSC Code:
51111800
NACRES:
NA.77

€168.00


Estimated to ship on21 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

solid

potency

9 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

10% acetic acid: 5 mg/mL

storage temp.

2-8°C

General description

A cell-permeable biscarboxamidine derivative that acts as a selective inhibitor of protein arginine methyltransferase 1 (PRMT1; IC50 = 9.5 µM). Exhibits about 10-fold greater selectivity over PRMT5 (IC50 = 110 µM) and PRMT6 (IC50 = 118 µM). Shown to block BMP4-induced phosphorylation and activation of Smad1/Smad5 and asymmetric dimethylation of Arg3 on histone 4 (H4R3) in HaCaT cells transfected with PRMT1.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Primary Target
PRMT1
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Xu, J., et al, 2013. Mol. Cell.51, 1.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service