Skip to Content
Merck
All Photos(1)

Key Documents

SML1227

Sigma-Aldrich

Tavaborole

≥95% (HPLC)

Synonym(s):

1,3-Dihydro-5-fluoro-1-hydroxy-2,1-benzoxaborole, 5-Fluoro-1,3-dihydro-1-hydroxy-2,1-benzoxaborole, AN-2690, AN2690

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€112.00
50 MG
€341.00

€112.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€112.00
50 MG
€341.00

About This Item

Empirical Formula (Hill Notation):
C7H6BFO2
CAS Number:
Molecular Weight:
151.93
UNSPSC Code:
12352200
NACRES:
NA.77

€112.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

B1(OCc2c1ccc(c2)F)O

InChI

1S/C7H6BFO2/c9-6-1-2-7-5(3-6)4-11-8(7)10/h1-3,10H,4H2

InChI key

LFQDNHWZDQTITF-UHFFFAOYSA-N

General description

Tavaborole is a boron-based pharmaceutical agent. It has broad-spectrum oxaborole antifungal activity. Due to its low molecular weight, it facilitates maximal nail plate penetration than its predecessors.[1][2][3]

Biochem/physiol Actions

Tavaborole (AN2690) is a potent antifungal that targets the post-transfer editing site of leucyl-tRNA synthetase (LeuRS).
Tavaborole (AN2690) is a potent antifungal that targets the post-transfer editing site of leucyl-tRNA synthetase (LeuRS). Tavaborole forms a covalent adduct with the 3′ adenosine of tRNA(leu) at the editing site of fungal, but not bacterial LeuRS, locking the enzyme in an inactive conformation. Tavaborole was recently approved for the treatment of onychomycosis of the toenail in adults.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tavaborole for the treatment of onychomycosis
Elewski BE and Tosti A
Expert Opinion on Pharmacotherapy, 15(10), 1439-1448 (2014)
Spotlight on tavaborole for the treatment of onychomycosis
Jinna S and Finch J
Drug design, development and therapy, 9, 6185-6185 (2015)
The efficacy and safety of tavaborole, a novel, boron-based pharmaceutical agent: phase 2 studies conducted for the topical treatment of toenail onychomycosis.
Toledo-Bahena ME, et al.
J. Drugs Dermatol., 13(9), 1124-1132 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service