Skip to Content
Merck
All Photos(1)

Key Documents

218697

Sigma-Aldrich

Casein Kinase II Inhibitor I

The Casein Kinase II Inhibitor I, also referenced under CAS 17374-26-4, controls the biological activity of Casein Kinase II. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Casein Kinase II Inhibitor I, TBB, TBBt, 4,5,6,7-Tetrabromobenzotriazole, 4,5,6,7-Tetrabromo-2-azabenzimidazole, CK2 Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€150.00

€150.00


Estimated to ship on21 May 2025



Select a Size

Change View
10 MG
€150.00

About This Item

Empirical Formula (Hill Notation):
C6HBr4N3
CAS Number:
Molecular Weight:
434.71
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€150.00


Estimated to ship on21 May 2025


Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Brc1c2n[nH]nc2c(c(c1Br)Br)Br

InChI

1S/C6HBr4N3/c7-1-2(8)4(10)6-5(3(1)9)11-13-12-6/h(H,11,12,13)

InChI key

OMZYUVOATZSGJY-UHFFFAOYSA-N

General description

A cell-permeable benzotriazolo compound that acts as a higly selective, reversible, and ATP/GTP-competitive inhibitor of casein kinase-2 (CK2) (IC50 = 900 nM and 1.6 M, using rat liver and human recombinant CK2, respectively) and DYRK (IC50<1 M for DYRK1a). Its specificity has been demonstrated using a panel of 33 kinases, including PKA, PKB, PKC, MKK1, Erk2, p38, JNK, Lck, Lyn, and Syk. Also shown to inhibit NS3-associated viral helicase activities in HCV (IC50 = 20 and 60 M, using DNA and RNA substrate, respectively) and WNV (IC50 = 1.7 and 900 nM, using DNA and RNA substrate, respectively). A 10 mM (5 mg/1.15 ml) solution of Casein Kinase II Inhibitor I in DMSO, is also available.

Biochem/physiol Actions

Cell permeable: yes
Product competes with ATP.
Reversible: yes
Target IC50: 900 nM and 1.6 µM, using rat liver and human recombinant CK2, respectively; <1 µM for DYRK1a

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Other Notes

Borowski, P., et al. 2003. Eur. J. Biochem.270, 1645.
Sarno, S., et al. 2003. Biochem. J.374, 639.
Zien, P., et al. 2003. Biochem. Biophys. Res. Commun.306, 129.
Ruzzene, M., et al. 2002. Biochem. J.364, 41.
Sarno, S., et al. 2001. FEBS Lett.496, 44.
Szyszka, R., et al. 1995. Biochem. Biophys. Res. Commun.208, 418.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clairine I S Larsen et al.
Viruses, 15(6) (2023-06-28)
Minute Virus of Mice (MVM) is an autonomous parvovirus of the Parvoviridae family that replicates in mouse cells and transformed human cells. MVM genomes localize to cellular sites of DNA damage with the help of their essential non-structural phosphoprotein NS1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service