Skip to Content
Merck
All Photos(1)

Key Documents

PZ0037

Sigma-Aldrich

Crisaborole

≥98% (HPLC)

Synonym(s):

4-[(1,3-Dihydro-1-hydroxy-2,1-benzoxaborol-5-yl)oxy]benzonitrile, 5-(4-Cyanophenoxy)-1,3-dihydro-1-hydroxy-[2,1]-benzoxaborole, 5-(4-Cyanophenoxy)-2,3-dihydro-1-hydroxy-2,1-benzoxaborole, AN2728

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€99.50
25 MG
€402.00

€99.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€99.50
25 MG
€402.00

About This Item

Empirical Formula (Hill Notation):
C14H10BNO3
CAS Number:
Molecular Weight:
251.05
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€99.50


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

room temp

SMILES string

OB1OCC2=C1C=CC(OC3=CC=C(C#N)C=C3)=C2

InChI

1S/C14H10BNO3/c16-8-10-1-3-12(4-2-10)19-13-5-6-14-11(7-13)9-18-15(14)17/h1-7,17H,9H2

InChI key

USZAGAREISWJDP-UHFFFAOYSA-N

Biochem/physiol Actions

Crisaborole is a non-steroidal anti-inflammatory phosphodiesterase 4 (PDE4) inhibitor.
Crisaborole is a non-steroidal anti-inflammatory phosphodiesterase 4 (PDE4) inhibitor. Crisaborole has an IC50 value of 490 nM for PDE4 with similar IC50 values for release of cytokines TNF-α, IL-2, and IFN-γ, and shows little inhibition against other PDE isozymes. Crisaborole has been approved as a topical treatment for atopic dermatitis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen Dong et al.
The Journal of pharmacology and experimental therapeutics, 358(3), 413-422 (2016-06-30)
Psoriasis and atopic dermatitis are skin diseases affecting millions of patients. Here, we characterize benzoxaborole phosphodiesterase (PDE)-4 inhibitors, a new topical class that has demonstrated therapeutic benefit for psoriasis and atopic dermatitis in phase 2 or phase 3 studies. Crisaborole
Tsutomu Akama et al.
Bioorganic & medicinal chemistry letters, 19(8), 2129-2132 (2009-03-24)
A series of phenoxy benzoxaboroles were synthesized and screened for their inhibitory activity against PDE4 and cytokine release. 5-(4-Cyanophenoxy)-2,3-dihydro-1-hydroxy-2,1-benzoxaborole (AN2728) showed potent activity both in vitro and in vivo. This compound is now in clinical development for the topical treatment

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service