R1530 is a multi-kinase inhibitor and an anti tumor agent.
R1530 is a multi-kinase inhibitor with activity against several kinases associated with tumor growth and angiogenesis, including VEGFR, FGFR, PDGFR, Chk2 and Cdk2. In tumor cells, R1530 inhibits tubulin polyerization and mitotic checkpoint function, resulting in polyploidy. The compound R1530 inhibits tumor growth and angiogenesis in animal models.
International journal of molecular sciences, 20(9) (2019-05-01)
Polo-like kinase 4 (PLK4) is a cell cycle-regulated protein kinase (PK) recruited at the centrosome in dividing cells. Its overexpression triggers centrosome amplification, which is associated with genetic instability and carcinogenesis. In previous work, we established that PLK4 is overexpressed
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 50(11), 1148-1159 (2015-07-21)
Performance of a two-stage biofiltration system was investigated for removal of styrene-acetone mixtures. High steady-state acetone loadings (above C(in)(Ac) = 0.5 g.m(-3) corresponding to the loadings > 34.5 g.m(-3).h(-1)) resulted in a significant inhibition of the system's performance in both
Questions
Reviews
★★★★★ No rating value