Skip to Content
Merck
All Photos(1)

Documents

L2420

Sigma-Aldrich

Monoclonal Anti-LRP1 antibody produced in mouse

~1.0 mg/mL, clone LRP1-11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-A2MR, Anti-APOER, Anti-APR, Anti-Apolipoprotein E receptor, Anti-Apolipoprotein receptor, Anti-CD91, Anti-LRP, Anti-Lipoprotein receptor-related protein, Anti-Low density lipoprotein receptor-related protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

LRP1-11, monoclonal

form

buffered aqueous solution

mol wt

antigen ~500 kDa

species reactivity

human, rat, mouse

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: suitable
western blot: 1-2 μg/mL

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Monoclonal Anti-LRP1 (mouse IgG1 isotype) is derived from the hybridoma LRP1-11 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide corresponding to a fragment of human LRP1, conjugated to KLH. LDL receptor family. Low-density lipoprotein (LDL) receptor related protein 1 (LRP1) gene is mapped to human chromosome 12q13.3. LRP1 protein, belongs to the LDL receptor family. LRP1, a single-chain 600 kDa type I transmembrane glycosylated precursor protein, is produced in the endoplasmic reticulum. This protein is proteolytically cleaved in the Golgi complex into two subunits including 85-kDa membrane-bound C-terminal membrane spanning subunit and a noncovalently attached 515-kDa N-terminal subunit.

Specificity

Monoclonal Anti-LRP1 recognizes human, rat, and mouse LRP1.

Application

Monoclonal Anti-LRP1 antibody produced in mouse has been used in:
  • immunoblotting
  • enzyme linked immunosorbent assay
  • immunofluorescence

Biochem/physiol Actions

Low density lipoprotein receptor related protein 1 (LRP1) plays an essential role in early stages of embryonic development. It aids in lipoprotein metabolism, proteinase regulation, activation of lysosomal enzymes and cellular entry of viruses and toxins. Additionally, It is also implicated in cell signaling pathways, cell migration, and neurodevelopment. LRP1 interacts with at least 30 different ligands including lipoproteins, proteinases, proteinase-inhibitor complexes, extra cellular matrix (ECM) proteins and various intracellular proteins. Furthermore, LDL receptor related protein 1 also has an ability to associate with bacterial toxins and viruses.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at –20 °C. For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze at –20 °C in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discard if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deficits in receptor-mediated endocytosis and recycling in cells from mice with Gpr107 locus disruption
Zhou G, et al.
Journal of Cell Science, 127(18), 3916-3927 (2014)
TOPBP1 recruits TOP2A to ultra-fine anaphase bridges to aid in their resolution
Broderick R, et al.
Nature Communications, 6(1), 1-12 (2015)
Inflammatory mediators promote production of shed LRP1/CD91, which regulates cell signaling and cytokine expression by macrophages
Gorovoy M, et al.
Journal of Leukocyte Biology, 88(4), 769-778 (2010)
Beyond endocytosis: LRP function in cell migration, proliferation and vascular permeability
Lillis AP, et al.
Journal of Thrombosis and Haemostasis, 3(8), 1884-1893 (2005)
Identification of the low density lipoprotein (LDL) receptor-related protein-1 interactome in central nervous system myelin suggests a role in the clearance of necrotic cell debris
Fernandez-Castaneda A, et al.
The Journal of biological chemistry, 288(7), 4538-4548 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service