Skip to Content
Merck
All Photos(1)

Key Documents

SML1031

Sigma-Aldrich

Bioymifi

≥98% (HPLC)

Synonym(s):

(Z)-5-(5-[(3-[4-Bromophenyl]-2-imino-4-oxothiazolidin-5-ylidene)methyl]furan-2-yl)isoindoline-1,3-dione

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€77.70
25 MG
€305.00

€77.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€77.70
25 MG
€305.00

About This Item

Empirical Formula (Hill Notation):
C22H12BrN3O4S
CAS Number:
Molecular Weight:
494.32
UNSPSC Code:
12352200
NACRES:
NA.77

€77.70


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, orange to dark orange red

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

S1\C(=C/c3[o]c(cc3)c4cc5c(cc4)C(=O)NC5=O)\C(=O)N(C1=N)c2ccc(cc2)Br

InChI

1S/C22H12BrN3O4S/c23-12-2-4-13(5-3-12)26-21(29)18(31-22(26)24)10-14-6-8-17(30-14)11-1-7-15-16(9-11)20(28)25-19(15)27/h1-10,24H,(H,25,27,28)/b18-10-,24-22?

InChI key

ULBOWKXOFOTCMU-NLDKGBHCSA-N

Biochem/physiol Actions

Bioymifi induces apoptosis via the Tumor necrosis factor (TNF)-related apoptosis-inducing ligand (TRAIL) death receptor DR5 independent of TRAIL.
Bioymifi induces apoptosis via the Tumor necrosis factor (TNF)-related apoptosis-inducing ligand (TRAIL) death receptor DR5 independent of TRAIL. Bioymifi binds selectively to the extracellular domain of DR5 with little binding affinity to DR4, promoting DR5 aggregation and activation. Bioymifi caused apoptosis and cell death in a variety of human cancer cell lines including T98G human glioblastoma cells, lung cancer cell lines H460 and H1155, the cervical cancer cell line HeLa, the osteosarcoma cell line U2OS, the pancreatic carcinoma cell line Miapaca and the colon cancer cell line HT29.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Apoptosis regulation involves multiple pathways and molecules for cellular homeostasis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service