Skip to Content
Merck
All Photos(1)

Key Documents

444937

Sigma-Aldrich

MEK Inhibitor I

The MEK Inhibitor I, also referenced under CAS 297744-42-4, controls the biological activity of MEK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

MEK Inhibitor I

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€221.00

€221.00


Estimated to ship on18 May 2025



Select a Size

Change View
1 MG
€221.00

About This Item

Empirical Formula (Hill Notation):
C21H18N4OS
CAS Number:
Molecular Weight:
374.46
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€221.00


Estimated to ship on18 May 2025


Quality Level

Assay

≥95% (sum of two isomers, HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 10 mg/mL
methanol: 50 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable pyridine-containing vinylogous cyanamide compound that acts as a potent and selective inhibitor of MEK (IC50 = 12 nM) with little activity towards MKK3 and MKK4 (IC50 >1 µM). The inhibition is noncompetitive with respect to ERK and the compound displays significant affinity only towards ATP-bound MEK (i.e. noncompetitive with respect to ATP). Exhibits superior potency, solubility, and stability compared to U0126 (Cat. No. 662005) in aqueous solutions. Shown to protect against phorbol ester-mediated ear edema in mice (ED50 of 5 mg/kg, ip.).
A cell-permeable, potent, and selective inhibitor of MEK (IC50 = 12 nM) with little activity towards MKK3 and MKK4 (IC50 >1 µM). The inhibition is non-competitive with respect to ERK and the compound displays significant affinity only towards ATP-bound MEK (i.e. non-competitive with respect to ATP). Exhibits superior potency, solubility, and stability compared to U0126 (Cat. No. 662005) in aqueous solutions. Shown to protect against phorbol ester-mediated ear edema in mice (ED50 of 5 mg/kg, ip.).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
MEK
Product does not compete with ATP.
Reversible: no
Target IC50: 12 nM against MEK

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Wityak, J., et al. 2004. Bioorg. Med. Chem. Lett.14, 1483.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gretchen M Argast et al.
Cells, tissues, organs, 193(1-2), 114-132 (2010-11-03)
Epithelial to mesenchymal transition (EMT) plays a dual role in tumor progression. It enhances metastasis of tumor cells by increasing invasive capacity and promoting survival, and it decreases tumor cell sensitivity to epithelial cell-targeting agents such as epithelial growth factor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service