Skip to Content
Merck
All Photos(1)

Key Documents

SMB01056

Sigma-Aldrich

Farnesiferol C

≥90% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€376.00

€376.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€376.00

About This Item

Empirical Formula (Hill Notation):
C24H30O4
CAS Number:
Molecular Weight:
382.49
UNSPSC Code:
12352205
NACRES:
NA.25

€376.00


Check Cart for Availability

Request a Bulk Order

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

382.49

solubility

water: slightly soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1[C@@]2([C@@H](C([C@H]1CC2)(C)C)CC\C(=C\COc3cc4c(cc3)C=CC(=O)O4)\C)C

InChI

1S/C24H30O4/c1-16(5-9-20-23(2,3)21-11-13-24(20,4)28-21)12-14-26-18-8-6-17-7-10-22(25)27-19(17)15-18/h6-8,10,12,15,20-21H,5,9,11,13-14H2,1-4H3/b16-12+/t20-,21-,24+/m1/s1

InChI key

OCHZHKVSLMBEJP-QYQYHAIPSA-N

General description

Farnesiferol C is a sesquiterpene coumarin commonly derived from plants like Ferula sinkiangensis, Ferula asafoetida, and Ferula pseudalliacea. Current research indicates that this bioactive metabolite is an inhibitor and may possess diverse biological activities, including anticancer, antiangiogenic, antimutagenic, antiviral, and antitumor properties.

Application

Farnesiferol C is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Features and Benefits

  • Suitable for Biochemical and Biomedical research
  • Versatile and adaptable for wide variety of laboratory and research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jamal Kasaian et al.
Journal of Asian natural products research, 20(1), 27-35 (2017-09-28)
Farnesiferol C (FC) is a sesquiterpene coumarin, with a unique chemical structure, isolated from Ferula (Apiaceae) species including Ferula assa-foetida and Ferula szowitsiana. The compound is an important natural product with different biological activities. Recently, researchers have been interested in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service