Skip to Content
Merck

Skip To

SML0801

Palomid 529

≥98% (HPLC)

Synonym(s):

8-(1-Hydroxyethyl)-2-methoxy-3-((4-methoxybenzyl)oxy)-6H-benzo[c]chromen-6-one, 8-(1-Hydroxyethyl)-2-methoxy-3-[(4-methoxyphenyl)methoxy]-6H-dibenzo[b,d]pyran-6-one, P529, SG 00529

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C24H22O6
CAS Number:
Molecular Weight:
406.43
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

[o]1c2c(c4c([c]1=O)cc(cc4)C(O)C)cc(c(c2)OCc3ccc(cc3)OC)OC

InChI

1S/C24H22O6/c1-14(25)16-6-9-18-19-11-22(28-3)23(12-21(19)30-24(26)20(18)10-16)29-13-15-4-7-17(27-2)8-5-15/h4-12,14,25H,13H2,1-3H3

InChI key

YEAHTLOYHVWAKW-UHFFFAOYSA-N

Biochem/physiol Actions

Palomid 529 (P529) is a PI3K/Akt/mTOR inhibitor, which is unique in that it causes the dissociation of both the TORC1 and TORC2 complexes and inhibits both Akt and mTOR signaling. Palomid 529 has been shown to inhibit tumor growth, angiogenesis, and vascular permeability. Palomid 529 (P529) inhibited glioblastoma growth, penetrating the blood brain barrier with little affinity for ATP-binding cassette (ABC) drug efflux transporters ABCB1 and ABCG2. Palomid 529 has also shown promise as a possible treatment for macular degeneration and keloid disease.
Palomid 529 (P529) is a PI3K/Akt/mTPR inhibitor, which inhibits TORC1 and TORC2 complexes and both Akt and mTOR signaling.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML0775SML0230SML1310
assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: ≥4 mg/mL at warmed to 60 °C

solubility

DMSO: 10 mg/mL, clear

color

white to beige

color

white to light brown

color

white to beige

color

white to beige


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Articles

Discover Bioactive Small Molecules for Kinase Phosphatase Biology






Global Trade Item Number

SKUGTIN
SML0801-25MG04061832450964
SML0801-5MG04061837107160

Questions

Reviews

No rating value

Active Filters