Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C10H8BrN3O
CAS Number:
Molecular Weight:
266.09
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Pricing and availability is not currently available.
Quality Segment
assay
≥98% (HPLC)
form
powder
storage condition
desiccated
color
white to beige
solubility
DMSO: ≥5 mg/mL (warm DMSO)
storage temp.
room temp
SMILES string
NC1=NC(=O)C(Br)=C(N1)c2ccccc2
InChI
1S/C10H8BrN3O/c11-7-8(6-4-2-1-3-5-6)13-10(12)14-9(7)15/h1-5H,(H3,12,13,14,15)
InChI key
CIUUIPMOFZIWIZ-UHFFFAOYSA-N
Application
Bropirimine, an immunostimulating agent and toll like receptor (TLR7) agonist, is used in comparison with other TLR7 agonists such as imiquimod, R-848, loxoribine in interferon cell signaling research.[1] Bropirimine may be used in experimental autoimmune encephalomyelitis (EAE) and bladder cancer research.[2]
Biochem/physiol Actions
Bropirimine is an immunostimulant, anti-tumor agent
Bropirimine is an immunostimulating agent. The compound induces production of α and β interferons and enhances NK cell function. Bropirimine has antiproliferative effects in cancer cell lines and tumor growth in in vivo models.
signalword
Warning
hcodes
Hazard Classifications
Repr. 2
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
