Skip to Content
Merck
All Photos(1)

Key Documents

SMB01011

Sigma-Aldrich

Isoacteoside

≥90% (LC/MS-ELSD)

Synonym(s):

Isoacteoside A, Isoverbascoside

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€183.00

€183.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€183.00

About This Item

Empirical Formula (Hill Notation):
C29H36O15
CAS Number:
Molecular Weight:
624.59
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

€183.00


Check Cart for Availability

Request a Bulk Order

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

624.59

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1[C@H]([C@@H]([C@@H]([C@H]([C@@H]1C)O)O)O)O[C@@H]2[C@H]([C@@H](O[C@@H]([C@H]2O)COC(=O)\C=C\c4cc(c(cc4)O)O)OCCc3cc(c(cc3)O)O)O

InChI

1S/C29H36O15/c1-13-22(35)24(37)25(38)29(42-13)44-27-23(36)20(12-41-21(34)7-4-14-2-5-16(30)18(32)10-14)43-28(26(27)39)40-9-8-15-3-6-17(31)19(33)11-15/h2-7,10-11,13,20,22-33,35-39H,8-9,12H2,1H3/b7-4+/t13-,20+,22-,23+,24+,25+,26+,27-,28+,29-/m0/s1

InChI key

FNMHEHXNBNCPCI-QEOJJFGVSA-N

General description

Isoacteoside, also known as Isoverbascoside, is a phenylethanoid glycoside, a bioactive natural compound commonly derived from plants such as Pedicularis striata, Plantago australis, Paulownia coreana, Caryopteris incana, Acacia lasiopetala, Maytenus laevis, and Spergula japonica. Current research indicates that this plant metabolite is an inhibitor and may possess diverse biological activities, including anti-inflammatory, antioxidant, and neuroprotective properties.

Application

Isoacteoside is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

Isoacteoside exhibits significant inhibition of advanced glycation and product formation with IC50 values of 4.6-25.7 μM. It exerts anti-inflammatory, antidiaberic, antiviral (COVID-19) activity and cytoprotective action.

Features and Benefits

  • Suitable for Biochemical and Biomedical research
  • Versatile and adaptable for wide variety of laboratory and research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Son Tung Ngo et al.
Journal of chemical information and modeling, 60(12), 5771-5780 (2020-06-13)
The novel coronavirus (SARS-CoV-2) has infected several million people and caused thousands of deaths worldwide since December 2019. As the disease is spreading rapidly all over the world, it is urgent to find effective drugs to treat the virus. The
Chika Ifeanyi Chukwuma et al.
Journal of ethnopharmacology, 235, 329-360 (2019-02-16)
Diabetes and hypertension are pathophysiologically related diseases that co-exist with a wider complex of metabolic diseases having similar set of risk factors. There are numerous ethnopharmacological evidences on the anti-diabetic and/or anti-hypertensive properties of medicinal plants from various parts of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service