Skip to Content
Merck
All Photos(1)

Documents

PHL82615

Oroxylin A

phyproof® Reference Substance

Synonym(s):

5,7-Dihydroxy-6-methoxy-2-phenyl-4H-1-benzopyran-4-one, 5,7-Dihydroxy-6-methoxyflavone;, 6-Methoxybaicalein;

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12O5
CAS Number:
Molecular Weight:
284.26
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

SMILES string

COc1c(O)cc2OC(=CC(=O)c2c1O)c3ccccc3

InChI

1S/C16H12O5/c1-20-16-11(18)8-13-14(15(16)19)10(17)7-12(21-13)9-5-3-2-4-6-9/h2-8,18-19H,1H3

InChI key

LKOJGSWUMISDOF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Oroxylin A is a flavonoid isolated from Scutellaria baicalensis, which is one of the most important medicinal herbs in traditional Korean/Chinese/Japanese medicine. The amelioration of Abeta(25-35) peptide-induced memory impairment by oroxylin A is believed to be mediated via the GABAergic neurotransmitter system after a single administration, or by reductions in Abeta(25-35) peptide-induced astrocyte and microglia activations, iNOS expression, lipid peroxidation, and increased cholinergic neurotransmission after subchronic administration.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Kai et al.
Free radical biology & medicine, 153, 89-102 (2020-04-15)
It is well acknowledged that alcoholic liver disease (ALD) is widely prevalent all over the world, characterized by aberrant lipid deposition and excessive oxidative stress in hepatocytes. Recently, pyroptosis, a new type of programmed cell death, has been found in
Min Shen et al.
International immunopharmacology, 85, 106637-106637 (2020-06-09)
More and more evidence showed that autophagy is an inflammation-related defense mechanism against a variety of diseases including liver fibrosis. However, the essential mechanisms remain poorly understood. In this study, we sought to elucidate the impact of Oroxylin A on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service