Skip to Content
Merck

Skip To

SML2213

CPI-637

≥98% (HPLC)

Synonym(s):

(4R)-1,3,4,5-Tetrahydro-4-methyl-6-[1-methyl-3-(1-methyl-1H-pyrazol-4-yl)-1H-indazol-5-yl]-2H-1,5-benzodiazepin-2-one, (4R)-4-Methyl-6-[1-methyl-3-(1-methylpyrazol-4-yl)indazol-5-yl]-1,3,4,5-tetrahydro-1,5-benzodiazepin-2-one, 2H-1,5-Benzodiazepin-2-one, 1,3,4,5-tetrahydro-4-methyl-6-[1-methyl-3-(1-methyl-1H-pyrazol-4-yl)-1H-indazol-5-yl]-, (4R)-, CPI 637

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg
Check Cart for Availability
€ 302,00

About This Item

Empirical Formula (Hill Notation):
C22H22N6O
CAS Number:
Molecular Weight:
386.45
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder

€ 302,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -39 to -49°, c = 1.0 in chloroform-d

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[n]1(ncc(c1)c2n[n](c3c2cc(cc3)c4c5c(ccc4)NC(=O)C[C@H](N5)C)C)C

InChI

1S/C22H22N6O/c1-13-9-20(29)25-18-6-4-5-16(22(18)24-13)14-7-8-19-17(10-14)21(26-28(19)3)15-11-23-27(2)12-15/h4-8,10-13,24H,9H2,1-3H3,(H,25,29)/t13-/m1/s1

InChI key

BFTKDWYIRJGJCA-CYBMUJFWSA-N

Biochem/physiol Actions

CBP/p300 Bromodomain inhibitor
CPI-637 is a cell penetrant, selective and potent CBP/p300 bromodomain inhibitor.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML1781SML1333SML2384
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: 5 mg/mL, clear (warmed)

solubility

DMSO: 2 mg/mL, clear

color

white to beige

color

white to beige

color

white to beige

color

white to beige

optical activity

[α]/D -39 to -49°, c = 1.0 in chloroform-d

optical activity

-

optical activity

-

optical activity

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters