These concentrated serum replacements are defined, as multipurpose products designed to eliminate the need for serum in culture. These products do not contain growth factors, steroid hormones, or mitogens.
S0638 Serum Replacement 1, 50X
Total protein concentration at 1X of 770 mg/mL
Highly purified bovine serum albumin (heat treated)
Bovine transferrin (heat treated)
Bovine insulin
S9388 Serum Replacement 2, 50X
Total protein concentration at 1X of 570 mg/mL
Highly purified bovine serum albumin (heat treated)
Bovine transferrin (heat treated)
Bovine insulin
S2640 Serum Replacement 3, 50X
Total protein concentration at 1X of 664 mg/mL
Highly purified human serum albumin (heat treated)
Human transferrin (heat treated)
Human (recombinant) insulin
S9388
Serum Replacement 2 (50×)
liquid, sterile-filtered, suitable for cell culture
About This Item
Recommended Products
biological source
bovine
sterility
sterile-filtered
form
liquid
technique(s)
cell culture | mammalian: suitable
impurities
endotoxin, tested
storage temp.
2-8°C
Other Notes
Caution
related product
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
What is the difference between Serum Replacement 1, 2, and 3?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service