Skip to Content
Merck

Skip To

S2640

Serum Replacement 3 (50×)

liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Serum Substitute

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mL
Check Cart for Availability
€ 68,70
100 mL
Check Cart for Availability
€ 521,00

About This Item

NACRES:
NA.71
UNSPSC Code:
12352207
Sterility:
sterile-filtered
Form:
liquid

€ 68,70


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

sterile-filtered

Quality Segment

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

storage temp.

2-8°C

General description

Sigma′s Serum Replacement 3 is a fortified, multipurpose serum replacement for the long-term culturing of anchorage dependant and suspension cultures. This serum replacement is effective in research situations such as: studying the effects of growth factors or in the manufacturing of cell derived products.

Application

Serum Replacement 3 (50×) has been used as a supplement in modified phosphate-buffered saline (DPBS) and as a replacement for the growth medium for in vitro experiments for Tendon cells.

Analysis Note

Stable for 30 days when diluted in cell culture medium.

Other Notes

Defined serum replacement designed primarily for the long-term growth of human cells or other mammalian cells. Recommended for use in the production of cell-secreted proteins. Contains only human proteins (i.e., human serum albumin, human transferrin, human recombinant insulin). Does not contain growth factors, steroid hormones, glucocorticoids, cell adhesion factors, detectable Ig and mitogens. May be used with anchorage-dependent and suspension cells.

Disclaimer

This product is tested and found non-reactive for Hepatitis B Surface Antigen (HBsAg) and non-reactive for Human Immunodeficiency Virus (HIV) antibody by ELISA. Nevertheless, products of human origin should be considered potentially infectious and handled accordingly.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S9388H1270F7942
form

liquid

form

liquid

form

liquid

form

-

sterility

sterile-filtered

sterility

sterile-filtered

sterility

sterile-filtered

sterility

γ-irradiated, sterile-filtered

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | hybridoma: suitable, cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

impurities

endotoxin, tested

impurities

≤10 EU/mL Endotoxin

impurities

≤10 EU/mL Endotoxin

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

Quality Level

200

Quality Level

-

Quality Level

500

Quality Level

500


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. What is the difference between Serum Replacement 1, 2, and 3?

    1 answer
    1. These concentrated serum replacements are defined, as multipurpose products designed to eliminate the need for serum in culture. These products do not contain growth factors, steroid hormones, or mitogens.

      S0638 Serum Replacement 1, 50X
      Total protein concentration at 1X of 770 mg/mL
      Highly purified bovine serum albumin (heat treated)
      Bovine transferrin (heat treated)
      Bovine insulin

      S9388 Serum Replacement 2, 50X
      Total protein concentration at 1X of 570 mg/mL
      Highly purified bovine serum albumin (heat treated)
      Bovine transferrin (heat treated)
      Bovine insulin

      S2640 Serum Replacement 3, 50X
      Total protein concentration at 1X of 664 mg/mL
      Highly purified human serum albumin (heat treated)
      Human transferrin (heat treated)
      Human (recombinant) insulin

      Helpful?

  2. Does the Serum Replacement 3 contain heat-treated bovine serum albumin, transferrin and insulin, like with Serum Replacement 1 and 2?

    1 answer
    1. As described in the "Other Notes" section of the product detail page, this is a defined serum replacement designed primarily for the long-term growth of human cells or other mammalian cells. Recommended for use in the production of cell-secreted proteins. Contains only human proteins (i.e., human serum albumin, human transferrin, human recombinant insulin). Does not contain growth factors, steroid hormones, glucocorticoids, cell adhesion factors, detectable Ig and mitogens. May be used with anchorage-dependent and suspension cells. It does not contain bovine proteins like products S0638, Serum Replacement 1 (50×) and S9388, Serum Replacement 2 (50×).

      Helpful?

Reviews

No rating value

Active Filters