Skip to Content
Merck
All Photos(1)

Key Documents

14591C

SAFC

EX-CELL® HeLa Serum-Free Medium for HeLa Cells

Animal-protein free, without L-glutamine, liquid, suitable for cell culture

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
€83.00
1000 ML
€150.00

€83.00


Check Cart for Availability


Select a Size

Change View
500 ML
€83.00
1000 ML
€150.00

About This Item

MDL number:
UNSPSC Code:
12352207

€83.00


Check Cart for Availability

Quality Level

description

for research or for further manufacturing use
serum-free

sterility

sterile

form

liquid

technique(s)

cell culture | mammalian: suitable

components

glucose: 6.0 g/L
HEPES: no
L-glutamine: no
NaHCO3: 1.8 g/L

shipped in

ambient

storage temp.

2-8°C

Application

EX-CELL HeLa is an animal-protein free, serum-free liquid medium developed for the long-term growth of HeLa cells in suspension culture. The HeLa cell line is a clonal derivative of the parent HeLa cell line and is capable of growth in suspension culture. HeLa suspension cultures can be subcultured directly into EX-CELL HeLa from serum-supplemented or serum-free media. Suspension cultures in EX-CELL HeLa have been carried for more than 25 passages with no loss of growth or viability.

Legal Information

EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Parvaneh Katoli et al.
Protein expression and purification, 147, 38-48 (2018-02-24)
Myocilin (MYOC) is a secreted protein found in human aqueous humor (AH) and mutations in the MYOC gene are the most common mutation observed in glaucoma patients. Human AH analyzed under non-reducing conditions suggests that MYOC is not normally found

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service