Skip to Content
Merck

Skip To

S6948

SGK2, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

H-SGK2, dJ138B7.2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 μg
Check Cart for Availability
€ 385,00

About This Item

NACRES:
NA.32
UNSPSC Code:
12352200
Specific activity:
70-94 nmol/min·mg
Assay:
≥70% (SDS-PAGE)
Recombinant:
expressed in baculovirus infected Sf9 cells

€ 385,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in baculovirus infected Sf9 cells

Quality Segment

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

70-94 nmol/min·mg

mol wt

~71 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SGK2(10110)

Biochem/physiol Actions

SGK2 is a member of the serum- and glucocorticoid-induced kinases (SGK) which are serine-threonine kinases and belong to the "AGC" kinase subfamily, which includes protein kinases A, G, and C, and its catalytic domain is most similar to protein kinase B (PKB).SGK2, like the other two isoforms SGK1 and SGK3, is stimulated by insulin and insulin-like growth factor-1 (IGF-1), and has been shown to enhance Na(+)/K(+)-ATPase activity in a variety of cells.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S7570SAB1406925SAB1410063
assay

≥70% (SDS-PAGE)

assay

≥75% (SDS-PAGE)

assay

-

assay

-

specific activity

70-94 nmol/min·mg

specific activity

100 units/mg protein (Bradford)

specific activity

-

specific activity

-

Gene Information

human ... SGK2(10110)

Gene Information

human ... SGK2(10110)

Gene Information

human ... SGK2(10110)

Gene Information

human ... SGK2(10110)

recombinant

expressed in baculovirus infected Sf9 cells

recombinant

expressed in E. coli

recombinant

-

recombinant

-

form

buffered aqueous glycerol solution

form

aqueous glycerol solution

form

buffered aqueous solution

form

buffered aqueous solution

mol wt

~71 kDa

mol wt

protein apparent mol wt ~66 kDa

mol wt

antigen ~41.2 kDa

mol wt

antigen 41.2 kDa


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters