Skip to Content
Merck

Skip To

S0149

Streptolysin O from Streptococcus pyogenes

From Streptococcus pyogenes, Thiol-activated toxin, lyophilized powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
25000 units
Check Cart for Availability
€ 246,00
€ 209,10

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.77
EC Number:
308-500-3
MDL number:
Form:
lyophilized powder
Quality level:

€ 209,10

List Price€ 246,00Save 15%
Web-Only Promotion

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Streptolysin O from Streptococcus pyogenes, γ-irradiated

biological source

Streptococcus pyogenes

Quality Level

sterility

γ-irradiated

form

lyophilized powder

application(s)

cell analysis

storage temp.

2-8°C

Gene Information

Streptococcus pyogenes ... slo(900490)

Application

Permeabilizes membranes to permit cellular uptake of large or charged molecules.
Streptolysin O from Streptococcus pyogenes has been used in streptolysin O (SLO) permeabilization. It has also been used to evaluate the barrier integrity and cytotoxicity of caco-2 and T84 IEC polarized monolayers.

Biochem/physiol Actions

Thiol-activated toxin that permeabilizes animal cell membranes. The protein binds as a monomer to membrane cholesterol and subsequently polymerizes into large arc- and ring-shaped structures surrounding pores of >12 nm.
Thiol-activated toxin that permeabilizes animal cell membranes..
Streptolysin O (SLO), obtained from Streptococcus pyogenes has the capability to block osteoclastogenesis, induced by receptor activator of nuclear factor κ-Β ligand (RANK) through the NF-κB signaling pathway. Streptococcus pyogenes (GAS) is a bacterial strain that results in bacterial arthritis, osteomyelitis and implant infections.

Preparation Note

Prepared from S 5265

Other Notes

One unit will cause 50% lysis of 2% red blood cell suspensions in phosphate buffered saline, pH 7.4, after incubation at 37 °C for 30 min.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S5265SAE0089D0564
form

lyophilized powder

form

lyophilized powder

form

-

form

lyophilized powder

Quality Level

200

Quality Level

200

Quality Level

-

Quality Level

200

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

application(s)

cell analysis

application(s)

-

application(s)

-

application(s)

-

Gene Information

Streptococcus pyogenes ... slo(900490)

Gene Information

Streptococcus pyogenes MGAS8232 ... slo(993428)

Gene Information

-

Gene Information

-

biological source

Streptococcus pyogenes

biological source

-

biological source

-

biological source

-


Still not finding the right product?


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 1 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Instructions


Elevated levels of StAR-related lipid transfer protein 3 alter cholesterol balance and adhesiveness of breast cancer cells: potential mechanisms contributing to progression of HER2-positive breast cancers
Vassilev B, et al.
The American Journal of Pathology, 185(4), 987-1000 (2015)
Streptolysin O derived from Streptococcus pyogenes inhibits RANKL-induced osteoclastogenesis through the NF-kappaB signaling pathway
Yi J, et al.
Molecular Medicine Reports (2018)
Extended exposure duration of cultured intestinal epithelial cell monolayers in characterizing hazardous and non-hazardous proteins
Zimmermann C, et al.
Food And Chemical Toxicology, 115(4), 451-459 (2018)



Global Trade Item Number

SKUGTIN
S0149-25KU04061836830083

Questions

Reviews

No rating value

Active Filters