Skip to Content
Merck

Skip To

RAB0844

Mouse Tnfrsf13B / Tumor Necrosis Factor Receptor Superfamily Member 13B ELISA Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
96 wells
Usually Ships in 4 Weeks. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
€ 848,00

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
mouse ... Tnfrsf13B(57916)
Species reactivity:
mouse
Storage temp.:
−20°C
Shipped in:
wet ice

€ 848,00


Usually Ships in 4 Weeks. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

assay range

inter-assay cv: <10%
intra-assay cv: <12%

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Tnfrsf13B(57916)

General description

The antibody pair provided in this kit recognizes mouse Tumor Necrosis Factor Receptor Superfamily Member 13B.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RAB0760RAB0788RAB0758
species reactivity

mouse

species reactivity

mouse

species reactivity

mouse

species reactivity

mouse

Gene Information

mouse ... Tnfrsf13B(57916)

Gene Information

mouse ... Tnfrsf17(21935)

Gene Information

mouse ... Tnfrsf18(21936)

Gene Information

mouse ... Tnfrsf13C(72049)

assay range

inter-assay cv: <10%
intra-assay cv: <12%

assay range

inter-assay cv: <10%
intra-assay cv: <12%

assay range

inter-assay cv: <10%
intra-assay cv: <12%

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 28 pg/mL
standard curve range: 28.67-7000 pg/mL

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)
    SDS

  • ELISA Stop Solution (Item I)
    SDS

  • 20X Wash Buffer (Item B)
    SDS

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
RAB0844-1KT04061835166206

Questions

Reviews

No rating value

Active Filters