Skip to Content
Merck

Skip To

RAB0635

Human CTSL / Cathepsin L1 ELISA Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
96 wells
Usually Ships in 4 Weeks. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
€ 771,00

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
human ... CTSL(1514)
Input:
sample type cell culture supernatant(s)
sample type plasma
sample type serum
Species reactivity:
human
Storage temp.:
−20°C
Shipped in:
wet ice

€ 771,00


Usually Ships in 4 Weeks. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 24 pg/mL
standard curve range: 24.58-6000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CTSL(1514)

General description

The antibody pair provided in this kit recognizes human Cathepsin L.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RAB0111RAB0737RAB1317
Gene Information

human ... CTSL(1514)

Gene Information

human ... CTSS(1520)

Gene Information

human ... TNFRSF13B(23495)

Gene Information

human ... CTSD(1509)

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)
sample type urine

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 24 pg/mL
standard curve range: 24.58-6000 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 4 pg/mL
standard curve range: 4.1-1000 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.3 ng/mL
standard curve range: 0.328-80 ng/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 50 pg/mL
standard curve range: 0.049-12 ng/mL

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)
    SDS

  • ELISA Stop Solution (Item I)
    SDS

  • 20X Wash Buffer (Item B)
    SDS

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content






Global Trade Item Number

SKUGTIN
RAB0635-1KT04061835164295

Questions

Reviews

No rating value

Active Filters