An anti-pan-protein-specific antibody will recognize the specific protein regardless of phosphorylation state (or other possible post-translational modification). An anti-phospho-protein specific antibody will recognize the protein only in a phosphorylated state.
Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 kit | Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery) | € 1.250,00 |
About This Item
mouse ... MAPK1(26413), MAPK14(26416), MAPK3(26417), MAPK8(26419)
rat ... MAPK1(116590), MAPK14(81649), MAPK3(50689), MAPK8(116554)
€ 1.250,00
usage
192 assays
species reactivity
mouse, human, rat
technique(s)
ELISA: suitable, capture ELISA: suitable
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... MAPK1(5594), MAPK14(1432), MAPK3(5595), MAPK8(5599)
mouse ... MAPK1(26413), MAPK14(26416), MAPK3(26417), MAPK8(26419)
rat ... MAPK1(116590), MAPK14(81649), MAPK3(50689), MAPK8(116554)
General description
Immunogen
Application
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
1 of 1
This Item | |||
|---|---|---|---|
| description 192 assays | description 96 assays | description 96 assays | description 96 assays |
| Gene Information human ... MAPK1(5594), MAPK14(1432), MAPK3(5595), MAPK8(5599) | Gene Information human ... MAPK1(5594), MAPK3(5595) | Gene Information human ... MAPK14(1432) | Gene Information human ... MAPK8(5599) |
| species reactivity mouse, human, rat | species reactivity rat, human, mouse | species reactivity human, rat, mouse | species reactivity human, rat, mouse |
| technique(s) ELISA: suitable, capture ELISA: suitable | technique(s) ELISA: suitable, capture ELISA: suitable | technique(s) ELISA: suitable, capture ELISA: suitable | technique(s) ELISA: suitable, capture ELISA: suitable |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| usage 192 assays | usage 96 assays | usage 96 assays | usage 96 assays |
| shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
Kit Components Also Available Separately
signalword
Danger
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
target_organs
Respiratory system
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What does PAN means? What is the difference between PAN and PHOSPHO? This is not described in the protocol
1 answer-
Helpful?
-






