Skip to Content
Merck

Skip To

RABHRP2

HRP-conjugated Anti-Mouse IgG Concentrate (Item I2)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 μL
Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)
€ 48,00

About This Item

NACRES:
NA.32
UNSPSC Code:
12352203
Clone:
-
Species reactivity:
-
Application:
Citations:
5

€ 48,00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


antibody product type

primary antibodies

shipped in

wet ice

storage temp.

−20°C


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RABTYROERABHRP1AP309P
shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

antibody product type

primary antibodies

antibody product type

primary antibodies

antibody product type

primary antibodies

antibody product type

secondary antibodies


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters