Skip to Content
Merck

Skip To

RABSTOP1

Stop Solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
14 mL
Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)
€ 22,80

About This Item

NACRES:
NA.32
UNSPSC Code:
12161703

€ 22,80


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


shipped in

wet ice

storage temp.

−20°C

Other Notes

Intended for use with ELISA kits.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GE18111250S-144A5852
storage temp.

−20°C

storage temp.

-

storage temp.

-10 to -25°C

storage temp.

2-8°C

shipped in

wet ice

shipped in

-

shipped in

-

shipped in

-


Still not finding the right product?

Explore all of our products under Stop Solution


pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters