Skip to Content
Merck

Skip To

RAB0189

Human FGF-9 ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 kit
Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)
€ 545,00

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
human ... FGF9(2254)
Input:
sample type urine
sample type serum
sample type plasma
sample type cell culture supernatant(s)
Species reactivity:
human
Storage temp.:
−20°C
Shipped in:
wet ice

€ 545,00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable, capture ELISA: suitable

input

sample type urine
sample type serum
sample type plasma
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 200 pg/mL
standard curve range: 358.4-35000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FGF9(2254)

General description

The Human FGF-9 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human FGF-9 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human FGF9

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RAB0187RAB0188RAB0233
species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

Gene Information

human ... FGF9(2254)

Gene Information

human ... FGF6(2251)

Gene Information

human ... FGF7(2252)

Gene Information

human ... IGFBP2(3485)

input

sample type urine
sample type serum
sample type plasma
sample type cell culture supernatant(s)

input

sample type cell culture supernatant(s)
sample type urine
sample type plasma
sample type serum

input

sample type cell culture supernatant(s)
sample type plasma
sample type urine
sample type serum

input

sample type cell culture supernatant(s)
sample type urine
sample type serum
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 200 pg/mL
standard curve range: 358.4-35000 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 6 pg/mL
standard curve range: 8.23-6000 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.5 pg/mL
standard curve range: 0.55-400 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 20 pg/mL
standard curve range: 8.23-6000 pg/mL

technique(s)

ELISA: suitable, capture ELISA: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable

packaging

kit of 96 wells (12 strips x 8 wells)

packaging

kit of 96 wells (12 strips x 8 wells)

packaging

kit of 96 wells (12 strips x 8 wells)

packaging

kit of 96 wells (12 strips x 8 wells)


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • ELISA 1X Assay/Sample Diluent Buffer A (Item D1)
    SDS

  • ELISA 5X Assay/Sample Diluent Buffer B (Item E1)
    SDS

  • ELISA Stop Solution (Item I)
    SDS

  • ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)
    SDS

  • 20X Wash Buffer (Item B)
    SDS

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content






Global Trade Item Number

SKUGTIN
RAB0189-1KT04061834960485

Questions

Reviews

No rating value

Active Filters