Skip to Content
Merck
All Photos(1)

Key Documents

Q4990

Sigma-Aldrich

QBP1

≥95% (HPLC)

Synonym(s):

L-Seryl-L-asparaginyl-L-tryptophyl-L-lysyl-L-tryptophyl-L-tryptophyl-L-prolylglycyl-L-isoleucyl-L-phenylalanyl-L-aspartic acid trifluoroacetate salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€218.00

€218.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€218.00

About This Item

Empirical Formula (Hill Notation):
C72H90N16O16 · xC2HF3O2
CAS Number:
Molecular Weight:
1435.58 (free base basis)
UNSPSC Code:
12352203
PubChem Substance ID:
NACRES:
NA.25

€218.00


Estimated to ship on01 June 2025


Request a Bulk Order

Assay

≥95% (HPLC)

form

powder

color

white

solubility

H2O: >1 mg/mL

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.CC[C@H](C)[C@H](NC(=O)CNC(=O)[C@@H]1CCCN1C(=O)[C@H](Cc2c[nH]c3ccccc23)NC(=O)[C@H](Cc4c[nH]c5ccccc45)NC(=O)[C@H](CCCCN)NC(=O)[C@H](Cc6c[nH]c7ccccc67)NC(=O)[C@H](CC(N)=O)NC(=O)[C@@H](N)CO)C(=O)N[C@@H](Cc8ccccc8)C(=O)N[C@@H](CC(O)=O)C(O)=O

InChI

1S/C72H90N16O16.C2HF3O2/c1-3-39(2)62(70(101)84-52(28-40-16-5-4-6-17-40)65(96)86-57(72(103)104)33-61(92)93)87-60(91)37-79-69(100)58-25-15-27-88(58)71(102)56(31-43-36-78-50-23-12-9-20-46(43)50)85-67(98)54(30-42-35-77-49-22-11-8-19-45(42)49)82-64(95)51(24-13-14-26-73)80-66(97)53(29-41-34-76-48-21-10-7-18-44(41)48)83-68(99)55(32-59(75)90)81-63(94)47(74)38-89;3-2(4,5)1(6)7/h4-12,16-23,34-36,39,47,51-58,62,76-78,89H,3,13-15,24-33,37-38,73-74H2,1-2H3,(H2,75,90)(H,79,100)(H,80,97)(H,81,94)(H,82,95)(H,83,99)(H,84,101)(H,85,98)(H,86,96)(H,87,91)(H,92,93)(H,103,104);(H,6,7)/t39-,47-,51-,52-,53-,54-,55-,56-,57-,58-,62-;/m0./s1

InChI key

VSUBSHMCMRUEAG-IQVZWRNHSA-N

Application

QBP1, an oligopeptide inhibitor of polyglutamine protein aggregation, may be used to study the physicochemical processes of polyglutamine-dependent protein misfolding, aggregation and precipitation. QBP1 may be used to study the role polyglutamine protein aggregation in causing or aggravating polyglutamine diseases including neurodegenerative diseases such as Huntington′s dentatorubral pallidoluysian atrophy, spinobulbar muscular atrophy, and spinocerebellar ataxia.[1][2]

Biochem/physiol Actions

QBP1 is an inhibitor of polyglutamine protein aggregation and cell death.
QBP1 is an inhibitor of polyglutamine protein aggregation and cell death. QBP1 inhibits polyglutamine aggregation in COS-7 cells at a concentration of 25 μM, as shown by complete inhibition of thioredoxin-Q62 aggregation assayed by turbidity at 405 nm. QBP1 reduces cell death of these cells by 50% and increases median life span from 5.5-52 days in Drosophilla melanogaster that expresses the expanded polyglutamine; can be shortened to 8 amino acids (Trp-Ley-Trp-Trp-Pro-Gly-Ile-Phe) without the loss of ability to inhibit polyglutamine aggregation. Several inherited neurodegenerative diseases, Huntington′s dentatorubral pallidoluysian atrophy, spinobulbar muscular atrophy, and spinocerebellar ataxia are caused by the expanded CAG repeats in the coding region of the gene, leading to accumulation of polyglutamine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miguel Mompeán et al.
Archives of biochemistry and biophysics, 675, 108113-108113 (2019-10-01)
Transactive Response DNA-Binding Protein of 43 kDa (TDP-43) is an essential human protein implicated in Amyotrophic Lateral Sclerosis (ALS) and common dementias. Its C-terminal disordered region, composed of residues 264-414 includes a hydrophobic segment (residues 320-340), which drives physiological liquid/liquid phase
Amit Mishra et al.
The Journal of biological chemistry, 283(12), 7648-7656 (2008-01-19)
The accumulation of intracellular protein deposits as inclusion bodies is the common pathological hallmark of most age-related neurodegenerative disorders including polyglutamine diseases. Appearance of aggregates of the misfolded mutant disease proteins suggest that cells are unable to efficiently degrade them
Peter O Bauer et al.
Nature biotechnology, 28(3), 256-263 (2010-03-02)
Huntington's Disease (HD) is a dominantly inherited pathology caused by the accumulation of mutant huntingtin protein (HTT) containing an expanded polyglutamine (polyQ) tract. As the polyglutamine binding peptide 1 (QBP1) is known to bind an expanded polyQ tract but not
H Akiko Popiel et al.
PloS one, 7(11), e51069-e51069 (2012-12-12)
The polyglutamine (polyQ) diseases such as Huntington's disease (HD), are neurodegenerative diseases caused by proteins with an expanded polyQ stretch, which misfold and aggregate, and eventually accumulate as inclusion bodies within neurons. Molecules that inhibit polyQ protein misfolding/aggregation, such as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service