Skip to Content
Merck
All Photos(1)

Key Documents

U5882

Sigma-Aldrich

U-74389G

Synonym(s):

21-[4-(2,6-Di-1-pyrrolidinyl-4-pyrimidinyl)-1-piperazinyl]pregna-1,4,9[11]-triene-3,20-dione maleate salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€270.00

€270.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 MG
€270.00

About This Item

Empirical Formula (Hill Notation):
C37H50N6O2 · C4H4O4
CAS Number:
Molecular Weight:
726.90
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€270.00


Estimated to ship on29 May 2025


Request a Bulk Order

solubility

ethanol: soluble 20 mg/mL, clear, light yellow

storage temp.

−20°C

SMILES string

OC(=O)\C=C/C(O)=O.[H][C@@]12CCC3=CC(=O)C=C[C@]3(C)C1=CC[C@]4(C)[C@H](CC[C@@]24[H])C(=O)CN5CCN(CC5)c6cc(nc(n6)N7CCCC7)N8CCCC8

InChI

1S/C37H50N6O2.C4H4O4/c1-36-13-11-27(44)23-26(36)7-8-28-29-9-10-31(37(29,2)14-12-30(28)36)32(45)25-40-19-21-42(22-20-40)34-24-33(41-15-3-4-16-41)38-35(39-34)43-17-5-6-18-43;5-3(6)1-2-4(7)8/h11-13,23-24,28-29,31H,3-10,14-22,25H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1-/t28-,29-,31+,36-,37-;/m0./s1

InChI key

ABCSSKWSUJMJCP-WQDFMEOSSA-N

Application

U-74389G has been shown to significantly decrease the level of ethanol-induced gastric mucosal necrosis in portal hypertensive (PHT) rats. Furthermore, U-74389G has been widely used in animal models of ischemic injury and hypertension[1].

Biochem/physiol Actions

Free radical lipid peroxidation inhibitor in cultured brain cells, monocytic THP1 cells, and LLC-PK1 cell layers.

Preparation Note

U-74389G is soluble in ethanol at 20 mg/ml and yields a clear, light yellow solution.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexios Bimpis et al.
Metabolic brain disease, 28(3), 439-446 (2013-01-25)
Spontaneous intracerebral hemorrhage (ICH) represents a partially-understood cerebrovascular disease of high incidence, morbidity and mortality. We, herein, report the findings of our study concerning the role of two important adenosinetriphosphatases (ATPases) in a porcine model of spontaneous ICH that we
Dimosthenis T Chrysikos et al.
The Journal of surgical research, 187(2), 450-457 (2013-12-18)
Oxidative stress is a crucial factor in the pathophysiology of acute pancreatitis and its systemic complications. Lazaroids are a novel class of antioxidants that potently protect pancreatic acinar cells against oxidant attack. The aim of our study was to evaluate
F Shi et al.
Free radical biology & medicine, 19(3), 349-357 (1995-09-01)
Selected 21-aminosteroids (U74500A, U74006F, and U74389G) and a 2-(aminomethyl)chromans (U78517F) were tested for their efficacy in preventing arachidonate-induced lipid peroxidation and permeability alterations in brain microvessel endothelial cells (BMECs). The 21-aminosteroids and 2-(aminomethyl)chromans were effective in varying degrees in inhibiting
H Kawanaka et al.
Hepatology (Baltimore, Md.), 34(5), 990-999 (2001-10-27)
Portal hypertensive (PHT) gastropathy is a frequent, serious complication of liver cirrhosis. PHT gastric mucosa has numerous abnormalities such as reduced mucosal potential differences, reduced surface oxygenation, and increased susceptibility to injury caused by alcohol, aspirin, and other noxious factors.
D J Crutchley et al.
Circulation, 92(2), 238-243 (1995-07-15)
Transition metals such as copper are known to initiate free radical formation and lipid peroxidation. Recent reports suggest that intracellular reactive oxygen intermediates can induce the transcription of a number of important genes. The present study examines the effects of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service