Skip to Content
Merck
All Photos(1)

Key Documents

SML1840

Sigma-Aldrich

YK-3-237

Synonym(s):

B-[2-methoxy-5-[(1E)-3-oxo-3-(3,4,5-trimethoxyphenyl)-1-propen-1-yl]phenyl]-boronic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€91.80

€91.80


Estimated to ship on02 June 2025



Select a Size

Change View
5 MG
€91.80

About This Item

Empirical Formula (Hill Notation):
C19H21BO7
CAS Number:
Molecular Weight:
372.18
UNSPSC Code:
12352200
NACRES:
NA.77

€91.80


Estimated to ship on02 June 2025


Assay

≥98% (HPLC)

Quality Level

form

powder

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

YK-3-237 is an activator of sirtuin-1 (SIRT1).
YK-3-237 is an activator of sirtuin-1 (SIRT1). YK-3-237 showed anti-proliferative activity toward triple-negative breast cancer, inducing G2/M cell cycle arrest and apoptosis. In a study on renal fibrosis, YK-3-237 caused enhanced expression of α-SMA and fibronectin, suggesting that long-term use of SIRT1 activators may increase the risk or progression of chronic kidney disease.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong Weon Yi et al.
Oncotarget, 4(7), 984-994 (2013-07-13)
Many types of mutations in tumor suppressor p53 are oncogenic through gain-of-function. Therefore, targeting mutant p53 (mtp53) is a promising therapeutic approach to fight against many types of cancers. We report here a small molecule compound YK-3-237 that reduces acetylation
Murugavel Ponnusamy et al.
The Journal of pharmacology and experimental therapeutics, 354(2), 142-151 (2015-05-30)
Although activation of sirtuin-1 (SIRT1) has been shown to protect the kidney from acute injury, its role in renal fibrosis remains controversial since both inhibition and activation of SIRT1 have been reported to attenuate renal fibrosis. To resolve this conflict

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service