Skip to Content
Merck
All Photos(1)

Key Documents

PZ0363

Sigma-Aldrich

CP-105,696

≥98% (HPLC)

Synonym(s):

(+)-1-(3S,4R)-[3-(4-Phenylbenzyl)-4-hydroxychroman-7-yl]cyclopentane carboxylic acid, 1-[(3S,4R)-3-([1,1′-Biphenyl]-4-ylmethyl)-3,4-dihydro-4-hydroxy-2H-1-benzopyran-7-yl]-cyclopentanecarboxylic acid, CP-105696, CP105696

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€144.00
25 MG
€444.00

€144.00


Estimated to ship on18 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€144.00
25 MG
€444.00

About This Item

Empirical Formula (Hill Notation):
C28H28O4
CAS Number:
Molecular Weight:
428.52
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

€144.00


Estimated to ship on18 April 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

storage temp.

room temp

SMILES string

OC(C1(CCCC1)C(C=C2)=CC3=C2[C@H](O)[C@@H](CC4=CC=C(C5=CC=CC=C5)C=C4)CO3)=O

InChI key

KMNLXCBYBKHKSK-BKMJKUGQSA-N

Application

CP-105,696 has been used as a leukotriene B4 receptor (LTB4R) antagonist to study the influence of the LTB4 pathway in the pathogenesis of acute itch flares in irritant calcipotriol (MC903)+ model allergen ovalbumin (OVA) sensitized murine model of atopic dermatitis (AD)-like disease.[1]

Biochem/physiol Actions

CP-105,696 is an orally active, potent and selective leukotriene B4 receptor (LTB4R) antagonist (IC50 against [3H]LTB4 = 5.6 nM for binding human neutrophils; IC50 = 9.7 nM & 30.3 nM, respectively using guinea pig spleen or mouse membranes) that targets LTB4R high- and low-affinity binding sites in a LTB4 non-competitive and competitive manner, respectively. CP-105,696 inhibits 5 nM LTB4-induced human neutrophils chemotaxis and CDllb upregualtion (IC50 = 5.2 nM and 430 nM, respectively) without inhibiting cyclooxygenase activity or affecting chemotaxis induced by C5a, PAF, IL-8. Oral administration is efficacious against intradermal LTB4 (100 ng/mouse), but not IL-1, injection-induced neutrophil accumulation in mice and guinea pigs (ED50 = 4.2 and 0.26 mgkg, respectively). CP-105,696 in vivo efficacy is also demonstrated in other animal models, including collagen-induced arthritis (CIA; 0.3-10 mg/kg in mice), allergic encephalomyelitis (ED50= 8.6 mg/kg; mice), and asthma (10-30 mg/kg; primate).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

(+)-1-(3S,4R)-[3-(4-phenylbenzyl)-4-hydroxychroman-7-yl]cyclopentane carboxylic acid, a highly potent, selective leukotriene B4 antagonist with oral activity in the murine collagen-induced arthritis model.
K Koch et al.
Journal of medicinal chemistry, 37(20), 3197-3199 (1994-09-30)
R J Griffiths et al.
Proceedings of the National Academy of Sciences of the United States of America, 92(2), 517-521 (1995-01-17)
Leukotriene B4 (LTB4) is a product of the 5-lipoxygenase pathway of arachidonic acid metabolism. LTB4 is a potent chemotactic factor for neutrophils and has been postulated to play an important role in a variety of pathological conditions including rheumatoid arthritis
C R Turner et al.
The Journal of clinical investigation, 97(2), 381-387 (1996-01-15)
To test the hypothesis that leukotriene (LT) B4 antagonists may be clinically useful in the treatment of asthma, CP-105,696 was evaluated in vitro, using chemotaxis and flow cytometry assays, and in vivo, using a primate asthma model. CP-105,696 inhibited LTB4-mediated
Fang Wang et al.
Cell, 184(2), 422-440 (2021-01-16)
Itch is an evolutionarily conserved sensation that facilitates expulsion of pathogens and noxious stimuli from the skin. However, in organ failure, cancer, and chronic inflammatory disorders such as atopic dermatitis (AD), itch becomes chronic, intractable, and debilitating. In addition to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service