Skip to Content
Merck
All Photos(1)

Key Documents

206700

Sigma-Aldrich

Calcein-AM

A cell-permeable, non-fluorescent, and hydrophobic compound, which is rapidly hydrolyzed by intracellular esterases releasing the membrane-impermeable, hydrophilic, and intensely fluorescent calcein.

Synonym(s):

Calcein-AM, Calcein-acetoxymethyl Ester, Diacetate, CAL-AM

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€301.00

€301.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€301.00

About This Item

Empirical Formula (Hill Notation):
C46H46N2O23
CAS Number:
Molecular Weight:
994.86
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.25

€301.00


Estimated to ship on23 May 2025


Request a Bulk Order

Quality Level

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 5 mg/mL

fluorescence

λex 494 nm
λem 517 nm

shipped in

ambient

storage temp.

−20°C

SMILES string

N(Cc1cc2c(cc1OC(=O)C)Oc3c(cc(c(c3)OC(=O)C)CN(CC(=O)OCOC(=O)C)CC(=O)OCOC(=O)C)C42OC(=O)c5c4cccc5)(CC(=O)OCOC(=O)C)CC(=O)OCOC(=O)C

InChI

1S/C46H46N2O23/c1-25(49)60-21-64-41(55)17-47(18-42(56)65-22-61-26(2)50)15-31-11-35-39(13-37(31)68-29(5)53)70-40-14-38(69-30(6)54)32(12-36(40)46(35)34-10-8-7-9-33(34)45(59)71-46)16-48(19-43(57)66-23-62-27(3)51)20-44(58)67-24-63-28(4)52/h7-14H,15-24H2,1-6H3

InChI key

BQRGNLJZBFXNCZ-UHFFFAOYSA-N

General description

A cell-permeable, non-fluorescent, and hydrophobic compound, which is rapidly hydrolyzed by intracellular esterases releasing the membrane-impermeable, hydrophilic, and intensely fluorescent calcein. Can also be used for imaging mitochondrial permeability pore transition. Calcein is better retained by viable cells than fluorescein, carboxyfluorescein, or BCECF, and its fluorescence is brighter in a variety of mammalian cell types. Useful for assessing adhesion, cell viability, chemotaxis, and multidrug resistance (MDR).
A cell-permeable, nonfluorescent, and hydrophobic compound, which is rapidly hydrolyzed by cytoplasmic esterases inside the cell, releasing the membrane-impermeable, hydrophilic, and intensely fluorescent calcein (excitation maximum: 494 nm; emission maximum: 517 nm). A cytosolic fluorophore that can also be used imaging mitochondrial permeability pore transition. Calcein is better retained by viable cells than fluorescein, carboxyfluorescein, and BCECF and its fluorescence is brighter in a variety of mammalian cell types. Useful for assessing adhesion, cell viability, chemotaxis and multidrug resistance (MDR).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Imaging mitochondrial permeability pore transition
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 4 months at -20°C.

Other Notes

Dhar, S., et al. 1998. Eur. J. Pharmacol. 346, 315.
Essodaigui, M., et al. 1998. Biochemistry 37, 2243.
Lemasters, J.J., et al. 1998. Biochim. Biophys. Acta 1366, 177.
Feller, N., et al. 1995. Br. J. Cancer 72, 543.
Feller, N., et al. 1995. FEBS Lett. 368, 385.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. ¿Condiciones de transporte y almacenamiento?

    1 answer
    1. This product stored at -20°C, but ships at ambient temperature. When there may be short-term exposure (i.e. during shipment) to conditions other than those recommended for long-term storage that will not affect product quality, Sigma-Aldrich will ship at ambient temperature.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service