Skip to Content
Merck
All Photos(1)

Key Documents

PZ0254

Sigma-Aldrich

PF-06273340

≥98% (HPLC)

Synonym(s):

N-(5-{[2-Amino-7-(2-hydroxy-1 ,1-dimethylethyl)-7H-pyrrolo[2,3-d]pyrimidin-5- yl]carbonyl}pyridin-3-yl)-2-(5-chloropyridin-2-yl)acetamide, N-[5-[[2-Amino-7-(2-hydroxy-1,1-dimethylethyl)-7H-pyrrolo[2,3-d]pyrimidin-5-yl]carbonyl]-3-pyridinyl]-5-chloro-2-pyridineacetamide, PF 06273340, PF06273340

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€124.00
25 MG
€501.00

€124.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€124.00
25 MG
€501.00

About This Item

Empirical Formula (Hill Notation):
C23H22ClN7O3
CAS Number:
Molecular Weight:
479.92
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

€124.00


Estimated to ship on30 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, white to yellow to brown

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

CC(CO)(C)N1C2=NC(N)=NC=C2C(C(C3=CN=CC(NC(CC4=CC=C(Cl)C=N4)=O)=C3)=O)=C1

InChI

1S/C23H22ClN7O3/c1-23(2,12-32)31-11-18(17-10-28-22(25)30-21(17)31)20(34)13-5-16(9-26-7-13)29-19(33)6-15-4-3-14(24)8-27-15/h3-5,7-11,32H,6,12H2,1-2H3,(H,29,33)(H2,25,28,30)

InChI key

BPIWZDNVMQQBQX-UHFFFAOYSA-N

Biochem/physiol Actions

PF-06273340 is a brain penetrant, orally available and potent tropomyosin-related kinase (Trk) inhibitor.
PF-06273340 is a brain penetrant, orally available and potent tropomyosin-related kinase (Trk) inhibitor. PF-06273340 is under development as a remedy acute and chronic pain.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service