Skip to Content
Merck
All Photos(1)

Key Documents

5.05823

Sigma-Aldrich

Galectin-1 Antagonist, DB21

Synonym(s):

Galectin-1 Antagonist, DB21, SVQNvaKL-[DBF]-IIVKLNA

Sign Into View Organizational & Contract Pricing

Select a Size

2 MG
€219.00

€219.00


Estimated to ship on15 May 2025



Select a Size

Change View
2 MG
€219.00

About This Item

Empirical Formula (Hill Notation):
C83H132N18O17
Molecular Weight:
1654.05
UNSPSC Code:
12352200
NACRES:
NA.77

€219.00


Estimated to ship on15 May 2025


Assay

≥98% (HPLC)

Quality Level

form

powder

potency

3 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 10 mg/mL

storage temp.

−20°C

General description

A dibenzofuran conjugated peptidomimetic (13-mer) that acts as an allosteric inhibitor of galectin-1 binding to cell surface glycans (IC50 = 3 µM). Shown to reduce the proliferation of endothelial cells and reduce vessel density in murine tumor models.

Biochem/physiol Actions

Primary Target
Galectin-1
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

SVQMKNva-[DBF]-IIVKLNA

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Dings, R.P., et al, 2013, J. Pharm. Exp. Ther.344, 589.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service