Skip to Content
Merck

Skip To

SML2216

Takinib

≥98% (HPLC), TAK1/MAP3K7 kinase inhibitor, powder

Synonym(s):

N1-(1-Propyl-1H-benzimidazol-2-yl)-1,3-benzenedicarboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
€ 100,00
25 mg
Check Cart for Availability
€ 405,00

About This Item

Empirical Formula (Hill Notation):
C18H18N4O2
CAS Number:
Molecular Weight:
322.36
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder

€ 100,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Takinib, ≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CCCN1C2=CC=CC=C2N/C1=N\C(C3=CC(C(N)=O)=CC=C3)=O

General description

Takinib belongs to the aminobenzimidazole class of inhibitors.

Application

Takinib has been used as an inhibitor of transforming 28 growth factor - β-activated kinase -1 (TAK-1) in human leukemia monocytic THP-1 cells. It may be used as a TAK1 inhibitor in Jurkat cells.

Biochem/physiol Actions

TAK-1 inhibitor
Takinib is a selective inhibitor of TAK1/MAP3K7 kinase, which is a member of the MAP3K family and a key regulator of cell death. It binds in the TAk1 ATPbinding pocket with an IC50 value of 9.5 nM. Takinib selectively induced apoptosis following TNF-α stimulation in rheumatoid arthritis and breast cancer.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML2825SML2050SML1762
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 20 mg/mL, clear

color

white to beige

color

white to beige

color

white to beige

color

, light yellow to dark orange


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters