Skip to Content
Merck

Skip To

PCRISPR006

Human CRISPR Poison Exon Knockout Library

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NE.02
UNSPSC Code:
41105904


packaging

pkg of 5 vials (5x200µL aliquots )

concentration

≥5x108 VP/ml (via p24 Assay)

application(s)

CRISPR

shipped in

dry ice

storage temp.

−70°C

General description

The human, paired guide Poison (pgPoison) library targets 3` splice sites with paired guide RNAs for alternative exon removal (pgRNAs), induce skipping of "poison" cassette exons and corresponding upstream constitutive exons in ultraconserved regions. This compact library (targets 963 exons) allows direct comparison between loss of a constitutive coding exon (knockout) with loss of a poison exon.

Custom pools for follow-up screening or 10x Genomics Compatible CRISPR pools are also available by contacting your local sales representative.

Application

  • Functional Genomics/Target Validation
  • Dual-gRNA
  • Poison exon
  • Internal controls included

Biochem/physiol Actions

In traditional CRISPR KO screens, Cas9 introduces double-strand breaks at locations specified by a gRNA. In this library, a CRISPR-Cas9-based method is used by expressing two sgRNAs to manipulate isoforms independent of gene inactivation. This approach enables rapid suppression of exon recognition in polyclonal settings to identify functional roles for individual exons.

Features and Benefits

  • Focus on your research, and we will generate your lentivirus screening library
  • Use CRISPR nucleases to knockout protein-coding genes to assess their function.
  • Built-in enrichment and depletion controls allow researchers to gauge the success of their pooled screening experiments confidently.
  • Dual-Guide Vector System (pools are gRNA-only, Cas9 sold separately) See products: LVCAS9BST or LVCAS9NEO.
  • Ease of optimization: Utilizes BFP and Puromycin as selection markers under EF1alpha promoter.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PCRISPR007PCRISPR008PCRISPR005
packaging

pkg of 5 vials (5x200µL aliquots )

packaging

pkg of 5 vials (5x200µL aliquots )

packaging

pkg of 5 vials (5x200µL aliquots )

packaging

pkg of 5 vials (5x200µL aliquots )

application(s)

CRISPR

application(s)

CRISPR

application(s)

CRISPR

application(s)

CRISPR

storage temp.

−70°C

storage temp.

−70°C

storage temp.

−70°C

storage temp.

−70°C

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

concentration

≥5x108 VP/ml (via p24 Assay)

concentration

≥5x108 VP/ml (via p24 Assay)

concentration

≥5x108 VP/ml (via p24 Assay)

concentration

≥5x108 VP/ml (via p24 Assay)


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



James D Thomas et al.
Nature genetics, 52(1), 84-94 (2020-01-09)
While RNA-seq has enabled comprehensive quantification of alternative splicing, no correspondingly high-throughput assay exists for functionally interrogating individual isoforms. We describe pgFARM (paired guide RNAs for alternative exon removal), a CRISPR-Cas9-based method to manipulate isoforms independent of gene inactivation. This



Global Trade Item Number

SKUGTIN
PCRISPR006-1EA04065266925289

Questions

Reviews

No rating value

Active Filters