Skip to Content
Merck

Skip To

PC0100

Protease Colorimetric Detection Kit

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352202


storage temp.

2-8°C

Application

Detection limit: 0.1 unit/ml (approx. 2 μg) trypsin with a 10 minute incubation time.
The Protease Colorimetric Detection Kit is designed for the accurate quantitation of protease activity. The method is based on the cleavage of casein into trichloroacetic acid-soluble peptides containing tyrosine and tryptophan residues. These peptides react with Folin-Ciocalteu reagent to give a characteristic blue color that can be measured at 660 nm.

Preparation Note

Supplied with sufficient reagents to perform 200 one mL assays.

Analysis Note

This kit has been tested for detection of all four proteolytic classes (serine, apartic, cysteine and metalloproteinases).

Other Notes

For greater sensitivity or for detection of trace protease activity, the Protease Fluorescent Detection kit (PF0100) is recommended.


Kit Components Only

Product No.
Description

  • Protease detection substrate (casein) 20 x 10

  • Folin & Ciocalteu's phenol reagent 10 mL

  • Sodium carbonate solution 125 mL

  • Enzyme diluent 125 mL

  • Tyrosine standard solution 1 mL

  • Protease from Bacillus polymyxa 1 g

  • Trichloroacetic acid (TCA) solution 5 mL



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library