Skip to Content
Merck

Skip To

P9768

Phorbol 12,13,20-triacetate

powder

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C26H34O9
CAS Number:
Molecular Weight:
490.54
UNSPSC Code:
12352202
PubChem Substance ID:
MDL number:


form

powder

color

white

storage temp.

−20°C

SMILES string

CC1C(OC(C)=O)C2(OC(C)=O)C(C3C=C(COC(C)=O)CC4(O)C(C=C(C)C4=O)C13O)C2(C)C

InChI

1S/C26H34O9/c1-12-8-19-24(31,21(12)30)10-17(11-33-14(3)27)9-18-20-23(6,7)26(20,35-16(5)29)22(34-15(4)28)13(2)25(18,19)32/h8-9,13,18-20,22,31-32H,10-11H2,1-7H3/t13-,18+,19-,20-,22-,24-,25-,26-/m1/s1

InChI key

LASMKIAVFGUYEG-SEYWVDNQSA-N



pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
P9768-5MG04061832865362

Questions

Reviews

No rating value

Active Filters