Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 10 g | Check Cart for Availability | € 44,60 € 37,91 |
About This Item
description
low viscosity
Quality Segment
form
powder
InChI
1S/C9H17NO5.Ca.2H/c1-9(2,5-11)7(14)8(15)10-4-3-6(12)13;;;/h7,11,14H,3-5H2,1-2H3,(H,10,15)(H,12,13);;;/t7-;;;/m0.../s1
InChI key
LPEKLWLORZBSDK-QTPLPEIMSA-N
Biochem/physiol Actions
A proprietary protein digest that provides increased stabilization of tissue sections in histochemical studies
Legal Information
Polypep is a registered trademark of Merck KGaA, Darmstadt, Germany
1 of 1
This Item | |||
|---|---|---|---|
| Quality Level 100 | Quality Level 200 | Quality Level 200 | Quality Level 100 |
| form powder | form powder | form - | form powder |
| description low viscosity | description - | description - | description - |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| P8736-100G | 04061833621929 |
| P8736-10G | 04061833621936 |
| P8736-50G | 04061833631478 |



