Skip to Content
Merck

Skip To

P7750

Peptone from animal tissue

from meat

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 g
Check Cart for Availability
€ 146,00
500 g
Check Cart for Availability
€ 368,00
1 kg
Check Cart for Availability
€ 674,00

About This Item

CAS Number:
UNSPSC Code:
41106212
eCl@ss:
42040102
NACRES:
NA.85
MDL number:

€ 146,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

animal (meat)

Quality Segment

form

powder

nitrogen analysis

≥10% total

loss

≤6% loss on drying

application(s)

microbiology

SMILES string

O(C)C(=O)C(CCC(CC)(CC)C)C(=O)OC

InChI

1S/C13H24O4/c1-6-13(3,7-2)9-8-10(11(14)16-4)12(15)17-5/h10H,6-9H2,1-5H3

InChI key

AIUDWMLXCFRVDR-UHFFFAOYSA-N

General description

Peptones are protein hydrolysates and widely used components of microbial media. Peptone serves as an organic nitrogen source for culturing microorganisms.[1]

Application

Peptone from animal tissue has been used:
  • to prepare neutralizing solution for viable cell count assay
  • to prepare decimal dilutions of the sample (cheese) for microbiological analyses[2]
  • in bolus liquid solution of amino acids to examine the effect of amino acid infusion in the stomach or intestine on specific dynamic action (SDA)[3]


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P590570173P4963
biological source

animal (meat)

biological source

animal (meat)

biological source

bovine milk

biological source

animal

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

application(s)

microbiology

application(s)

-

application(s)

food and beverages, microbiology

application(s)

food and beverages, microbiology

nitrogen analysis

≥10% total

nitrogen analysis

-

nitrogen analysis

-

nitrogen analysis

-

form

powder

form

powder

form

powder

form

powder

loss

≤6% loss on drying

loss

-

loss

≤10% loss on drying

loss

≤11% loss on drying


Still not finding the right product?

Explore all of our products under Peptone from animal tissue


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
P7750-500G04061834397182
P7750-1KG04061826764473
P7750-100G04061838591203

Questions

Reviews

No rating value

Active Filters