Skip to Content
Merck

Skip To

P5641

Polyoxyethylene (20) oleyl ether

Synonym(s):

Brij® 98, Brij® 99, C18-1E20

Sign In to View Organizational & Contract Pricing.

About This Item

CAS Number:
UNSPSC Code:
12161900
MDL number:


description

non-ionic

InChI

1S/C20H40O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-19-22-20-18-21/h9-10,21H,2-8,11-20H2,1H3/b10-9-

InChI key

KWVPFECTOKLOBL-KTKRTIGZSA-N

General description

Polyoxyethylene (20) oleyl ether is a non-ionic surfactant which can be used as structure-directing agents.

Application

Polyoxyethylene (20) oleyl ether has been used in a study to assess its effect on the transport of ibuprofen across rat skin. [1] It has also been used in a study to investigate novel potential carriers for non-invasive drug delivery. [2]

Legal Information

Brij is a registered trademark of Croda International PLC


pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

>464.0 °F - Equilibrium method

flash_point_c

> 240 °C - Equilibrium method

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Christian Wachter et al.
Journal of drug targeting, 16(7), 611-625 (2008-08-08)
Novel potential carriers for non-invasive drug delivery were prepared from polyoxyethylene(20) oleyl ether (C(18:1)EO(20)) and soybean phosphatidylcholine (SPC) in different relative molar ratios, R(e); this produced stiff SPC liposomes (2r(ves) approximately 120 nm) at one end and much smaller (2r(mic)
E S Park et al.
International journal of pharmaceutics, 209(1-2), 109-119 (2000-11-21)
The influence of polyethoxylated non-ionic surfactants on the transport of ibuprofen across rat skin was investigated. The skin permeation of ibuprofen from a series of 17 polyoxyethylene (POE) alkyl ethers containing 5% ibuprofen was determined using Franz diffusion cells fitted
Synthesis and photocatalytic activity of co-doped mesoporous TiO"2 on Brij98/CTAB composite surfactant template
Zhou, Z., et al.
Journal of Solid State Chemistry, 183, 6-6 (2010)