Skip to Content
Merck

Skip To

P4149

Poly(Glu, Ala, Tyr) sodium salt

Glu:Ala:Tyr (1:1:1), mol wt 20,000-50,000

Sign In to View Organizational & Contract Pricing.

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.26
MDL number:


form

solid

feed ratio

Glu:Ala:Tyr (1:1:1)

mol wt

20,000-50,000

storage temp.

−20°C

Analysis Note

Molecular weight based on viscosity.

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Articles

Humankind has utilized protein materials throughout its existence, starting with the use of materials such as wool and silk for warmth and protection from the elements and continuing with the use of recombinant DNA techniques to synthesize proteins with unique and useful properties.


S J Seo et al.
The Journal of investigative dermatology, 116(6), 880-885 (2001-06-16)
The physiologic role of gamma delta-T-cell-receptor-bearing cells and the T cell receptor ligands that they recognize is still poorly understood. Previous studies have suggested that one possible antigen for gamma delta-TCR(+) cells is the random copolymer poly-glutamic acid-tyrosine (poly-Glu-Tyr), because



Global Trade Item Number

SKUGTIN
P4149-25MG04061832779812
P4149-10MG04061832779805

Questions

Reviews

No rating value

Active Filters