Skip to Content
Merck

Skip To

P102

R(+)-3-(3-Hydroxyphenyl)-N-propylpiperidine hydrochloride

solid

Synonym(s):

R(+)-3-PPP hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
25 mg
Check Cart for Availability
€ 121,00
€ 102,85
100 mg
Check Cart for Availability
€ 299,00
€ 254,15

About This Item

Empirical Formula (Hill Notation):
C14H21NO · HCl
CAS Number:
Molecular Weight:
255.78
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Form:
solid

€ 102,85

List Price€ 121,00Save 15%
Web-Only Promotion

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solid

Quality Segment

color

white

solubility

H2O: 150 mg/mL, ethanol: slightly soluble

SMILES string

Cl.CCCN1CCC[C@@H](C1)c2cccc(O)c2

InChI

1S/C14H21NO.ClH/c1-2-8-15-9-4-6-13(11-15)12-5-3-7-14(16)10-12;/h3,5,7,10,13,16H,2,4,6,8-9,11H2,1H3;1H/t13-;/m0./s1

InChI key

NRHUDETYKUBQJT-ZOWNYOTGSA-N

Gene Information

human ... DRD2(1813)

Biochem/physiol Actions

D2 dopamine receptor agonist. Also functions as σ1 receptor antagonist with essentially no affinity for the phencylidine site on the NMDA receptor.

Preparation Note

Solutions should be freshly prepared.

Legal Information

Sold with the permission of Astra Pharmaceutical Co.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
54880D054H130
form

solid

form

solid

form

solid

form

solid

Quality Level

200

Quality Level

100

Quality Level

100

Quality Level

100

solubility

H2O: 150 mg/mL, ethanol: slightly soluble

solubility

H2O: 50 mg/mL, slightly hazy, faintly yellow

solubility

H2O: >5 mg/mL

solubility

H2O: soluble

color

white

color

-

color

white

color

white

Gene Information

human ... DRD2(1813)

Gene Information

-

Gene Information

human ... DRD1(1812)

Gene Information

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
P102-25MG04061832092508
P102-100MG04061832083551

Questions

Reviews

No rating value

Active Filters