Skip to Content
Merck

Skip To

OGS589

PBR322 - PBR322 LOW COPY CLONING VECTOR

plasmid vector for molecular cloning

Synonym(s):

cloning vector, expression vector, molecular cloning vector, plasmid, plasmid vector, snapfast vector, vector

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.85
UNSPSC Code:
12352200
Origin of replication:
BR322 (15 copies)
Bacteria selection:
ampicillin
Reporter gene:
none
Peptide cleavage:
no cleavage


form

buffered aqueous solution

mol wt

size 4361 bp

bacteria selection

ampicillin

origin of replication

BR322 (15 copies)

peptide cleavage

no cleavage

reporter gene

none

shipped in

ambient

storage temp.

−20°C

General description

This plasmid contains the BR322 (low copy number) origin of replication and provides approximately 20 plasmid copies per bacterial cell. It also contains the mammalian CMV promoter upstream of the MCS and Kan resistance for selection of transfected mammalian cells. Low copy number plasmids are useful for very large transgenes or for transgenes that can be toxic to bacterial cells.

Promoter Expression Level:

Application

BR322 Information: BR322 is a wild type plasmid isolated from E.coli. It contains a low copy origin that allows for plasmid maintenance at a frequency of approximately 10-100 plasmid copies per cell. Typically this number is closer to 30-40 copies per cell. Most common cloning plasmids contain a derivative of the origin of replication in this plasmid called pUC. The pUC origin was created by removing the Rep repressor protein that normally regulates plasmid copy number in E.coli and also making a single point mutation in the origin of replication itself. The point mutation is the most important difference between the origin of replication.This plasmid can be used as a general purpose low copy cloning vector.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.oxgene.com

Other Notes

To view sequence information for this product, please visit the product page


Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Geoffrey M Lynn et al.
Nature biotechnology, 33(11), 1201-1210 (2015-10-27)
The efficacy of vaccine adjuvants such as Toll-like receptor agonists (TLRa) can be improved through formulation and delivery approaches. Here, we attached small molecule TLR-7/8a to polymer scaffolds (polymer-TLR-7/8a) and evaluated how different physicochemical properties of the TLR-7/8a and polymer
Diana Romero et al.
Carcinogenesis, 37(1), 18-29 (2015-10-28)
Dickkopf-3 (Dkk-3) is a secreted protein whose expression is downregulated in many types of cancer. Endogenous Dkk-3 is required for formation of acini in 3D cultures of prostate epithelial cells, where it inhibits transforming growth factor (TGF)-β/Smad signaling. Here, we
Jin-Gyoung Jung et al.
PLoS genetics, 10(10), e1004751-e1004751 (2014-10-31)
The Notch3 signaling pathway is thought to play a critical role in cancer development, as evidenced by the Notch3 amplification and rearrangement observed in human cancers. However, the molecular mechanism by which Notch3 signaling contributes to tumorigenesis is largely unknown.



Global Trade Item Number

SKUGTIN
OGS589-5UG04061834249108

Questions

Reviews

No rating value

Active Filters