Skip to Content
Merck

Skip To

N8652

NF 023 hydrate

≥98% (HPLC), solid

Synonym(s):

8,8′-[Carbonylbis(imino-3,1-phenylene carbonylimino)]bis(1,3,5-naphthalene-trisulfonic acid) hexasodium salt hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Check Cart for Availability
€ 184,00
€ 156,40
50 mg
Check Cart for Availability
€ 593,00

About This Item

Empirical Formula (Hill Notation):
C35H20N4O21S6 · 6Na · xH2O
CAS Number:
Molecular Weight:
1162.88 (anhydrous basis)
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Storage condition:
desiccated

€ 156,40

List Price€ 184,00Save 15%
Web-Only Promotion

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

solid

storage condition

desiccated

color

white to off-white

solubility

H2O: >20 mg/mL

SMILES string

O.[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[O-]S(=O)(=O)c1cc(c2c(NC(=O)c3cccc(NC(=O)Nc4cccc(c4)C(=O)Nc5ccc(c6cc(cc(c56)S([O-])(=O)=O)S([O-])(=O)=O)S([O-])(=O)=O)c3)ccc(c2c1)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C35H26N4O21S6.6Na.H2O/c40-33(38-25-7-9-27(63(49,50)51)23-13-21(61(43,44)45)15-29(31(23)25)65(55,56)57)17-3-1-5-19(11-17)36-35(42)37-20-6-2-4-18(12-20)34(41)39-26-8-10-28(64(52,53)54)24-14-22(62(46,47)48)16-30(32(24)26)66(58,59)60;;;;;;;/h1-16H,(H,38,40)(H,39,41)(H2,36,37,42)(H,43,44,45)(H,46,47,48)(H,49,50,51)(H,52,53,54)(H,55,56,57)(H,58,59,60);;;;;;;1H2/q;6*+1;/p-6

InChI key

FLINBQMOKZJBEC-UHFFFAOYSA-H

Gene Information

Application

NF 023 hydrate has been used as an inhibitor of methyltransferases.[1] It has also been used to test its inhibitory effect on human adenovirus type-37 (HAdV-D37) viral infection in human corneal epithelial (HCE) cells.[2]

Biochem/physiol Actions

NF 023 contains six benzene rings and is a suramin analog.[2] It is a potent, competitive, and selective P2X purinoceptor antagonist[3] that aids in discriminating between P2X receptor subtypes.[3]

Features and Benefits

This compound is featured on the P2 Receptors: P2X Ion Channel Family page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Packaging

Desiccate

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
B3686P0021480415
assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥95% (HPLC)

form

solid

form

solid

form

powder

form

solid

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage condition

desiccated

storage condition

protect from light

storage condition

desiccated

storage condition

OK to freeze, desiccated (hygroscopic)

solubility

H2O: >20 mg/mL

solubility

H2O: >8 mg/mL

solubility

H2O: ≥20 mg/mL

solubility

water: soluble

color

white to off-white

color

white

color

white to off-white

color

white to off-white


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library