Skip to Content
Merck
All Photos(1)

Key Documents

N6000

Sigma-Aldrich

Naphthol AS-TR phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H15ClNO5P
CAS Number:
Molecular Weight:
391.74
Beilstein:
6666070
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

form

powder

storage temp.

−20°C

SMILES string

Cc1cc(Cl)ccc1NC(=O)c2cc3ccccc3cc2OP(O)(O)=O

InChI

1S/C18H15ClNO5P/c1-11-8-14(19)6-7-16(11)20-18(21)15-9-12-4-2-3-5-13(12)10-17(15)25-26(22,23)24/h2-10H,1H3,(H,20,21)(H2,22,23,24)

InChI key

SHTFMUPUBYXPCD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Substrate for the histochemical demonstration of acid and alkaline phosphatase.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander Apschner et al.
Disease models & mechanisms, 7(7), 811-822 (2014-06-08)
In recent years it has become clear that, mechanistically, biomineralization is a process that has to be actively inhibited as a default state. This inhibition must be released in a rigidly controlled manner in order for mineralization to occur in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service